STEMdiff™ Motor Neuron Differentiation Kit
Fournisseur: STEMCELL Technologies
100-0871
STMC100-0871EA 429 EUR InStock
STMC100-0871
STEMdiff™ Motor Neuron Differentiation Kit
Milieux de culture cellulaire
Differentiation kit for the generation of motor neurons from hPSCs.
With the STEMdiff™ motor neuron differentiation kit, you can produce pure populations of spinal motor neurons with an average of >80% Class III β-tubulin positive, >50% motor neuron marker ISL1 positive, >60% motor neuron marker HB9 positive. These motor neurons also exhibit high expression of HOXA5. Use the resulting motor neuron cultures in a variety of applications, such as modeling human neurological disease or toxicity testing. These highly versatile cultures are suitable for use in neuromuscular co-culture with myotubes generated using the MyoCult™ differentiation kit, in co-culture with microglia generated using the STEMdiff™ microglia differentiation kit, or with AggreWell™400 for higher yield production. For further maturation of motor neurons generated with this kit, pair with the STEMdiff™ motor neuron maturation kit to produce >40% ChAT cells in an additional 14 days.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité