STEMdiff™ Motor Neuron Maturation Kit
Fournisseur: STEMCELL Technologies
100-0872
STMC100-0872EA 399 EUR InStock
STMC100-0872
STEMdiff™ Motor Neuron Maturation Kit
Milieux de culture cellulaire
Differentiation and maturation kits for generation of motor neurons from human pluripotent stem cells.
Reduce the risk of culture failure by maturing and maintaining motor neuron cells using the STEMdiff™ Motor Neuron Maturation Kit. This kit combines neurophysiological BrainPhys™ Medium - proven to support long-term activity and maturation - with the supplements needed to produce functional motor neuron cultures. Cells matured with the STEMdiff™ Motor Neuron Maturation Kit exhibit high-level expression of expected motor neuron markers, including ChAT (>40%), across multiple cell lines. Use the resulting motor neuron cultures in a variety of applications, such as modeling human neurological disease or toxicity testing. The STEMdiff™ Motor Neuron Maturation Kit can also support co-culture with muscle cells generated using the MyoCult™ Differentiation Kit or with microglia generated using the STEMdiff™ Microglia Differentiation Kit.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité