Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Caspase-3/7 Activity Flow Cytometry Kit, Green

Fournisseur: STEMCELL Technologies

100-0923
STMC100-0923EA 562 EUR InStock
STMC100-0923
Caspase-3/7 Activity Flow Cytometry Kit, Green
Tests Tests cellulaires
For detection of active caspase-3 in mammalian cells undergoing apoptosis - 100 tests.

  • TF2-DEVD-FMK detects caspase-3/7 activity
  • Excitation wavelength: 503 nm
  • Emission wavelength: 525 nm
  • Extinction coefficient: 75,000 cm⁻¹M⁻¹
  • Easily scalable for high-throughput assays
  • Verified for use with flow cytometry

Efficiently detect active caspase-3 in mammalian cells undergoing apoptosis by using the Caspase-3/7 activity flow cytometry kit, green. This reagent kit uses a quick and simple staining protocol that is highly sensitive and compatible with standard flow cytometers.

Caspase-3 and caspase-7 have a proteolytic substrate selectivity for the peptide sequence Asp-Glu-Val-Asp (DEVD). This kit includes TF2-DEVD-FMK a cell-permeable and non-toxic fluorescent substrate as well as an assay buffer and propidium iodide. When added to apoptotic cells, TF2-DEVD-FMK irreversibly binds to activated caspase-3, resulting in the retention of the green fluorescent reagent within cells that can be detected by flow cytometry.

Caspases are a family of proteases that play a central role in cellular apoptosis through the cleavage of select proteins, and result in the disassembly of the cell (Thornberry & Lazebnik Science, 1998). This process is switched on by pro-apoptotic signals from death receptors or stimuli such as cytotoxic reagents, which activate the initiator caspases and lead to activation of the effector caspases, caspase-3 and caspase-7 (Thornberry & Lazebnik Science, 1998). Caspase-3/7 Activity Flow Cytometry Kit, Green is useful for screening caspase-3 inhibitors and quantifying apoptotic cells containing active caspase-3.

CAS No: 67-68-5

Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').


Commander maintenant

 

COMMANDE

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité