Human Recombinant Leptin
Fournisseur: STEMCELL Technologies
100-0932
STMC100-0932EA 319 EUR InStock
STMC100-0932
Human Recombinant Leptin
Protéines et Peptides
Leptin is a protein produced from the ob gene or lep gene, and acts as both a hormone involved in metabolic function as well as a proinflammatory cytokine by inducing Th1 immune responses (Dunn et al.; La Cava; Newman and Gonzalez-Perez). Leptin is a member of the gp130 family of cytokines and is known to be correlated with obesity (Dixit et al.; Dunn et al.). Leptin can both promote osteogenesis via osteoblast receptors and inhibit it through its actions on the hypothalamus (Figenschau et al.). It is a small, non-glycosylated protein with a highly conserved sequence between species. Leptin binds to the leptin receptor OB-R, which exists in six isoforms in humans and can activate various Janus kinase (JAK) and downstream pathways (Dunn et al.; Münzberg and Morrison; Newman and Gonzalez-Perez). For example, the binding of leptin to LepRb receptor activates JAK2, which leads to the phosphorylation of both JAK2 and the LepRb receptor, with three separate residues on the receptor triggering signaling pathways for SHP-2, STAT5, and STAT3 (Dunn et al.; Münzberg and Morrison). In addition to its effects on mature immune cells, other studies have suggested impacts on other cell types, with activities such as inducing cell proliferation and morphological differentiation in hematopoietic cell lines, chondrocytes, and hepatic cells (Figenschau et al.; Gainsford et al.; Santos-Alvarez et al.; Wang et al.).
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité