Human Recombinant R-Spondin-3, His Tag
Fournisseur: STEMCELL Technologies
100-0934
STMC100-0934EA 419 EUR InStock
STMC100-0934
Human Recombinant R-Spondin-3, His Tag
Protéines et Peptides
R-Spondin-3 is a member of thrombospondin type 1 repeat (TSR-1) superfamily that is involved in the canonical Wnt/β-catenin signaling pathway (de Lau et al.). R-spondin proteins are characterized by two furin-like repeats at the amino terminus and thrombospondin domain located near the carboxyl terminus (de Lau et al.). R-spondin-3 expression is associated with ovarian cancer (Gu et al.), prostate cancer (Mesci et al.), and differentiation of intestinal epithelial cells in diabetes mellitus (Shan et al.). In a transgenic mice model, the expression of R-Spondin-3 induces the expansion of Lgr5+ stem cells, Paneth cells, and Lgr4+ cells, promoting the intestinal stem cell compartment (Hilkens et al.). This protein contains a His-residue tag at the carboxyl end of the polypeptide chain.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité