Human Recombinant IL-1RA, ACF
Fournisseur: STEMCELL Technologies
100-0939
STMC100-0939EA 165 EUR InStock
STMC100-0939
Human Recombinant IL-1RA, ACF
Protéines et Peptides
Interleukin 1 receptor antagonist (IL-1RA) is a member of the IL-1 family that binds to IL-1 receptors but does not induce any intracellular signaling (Arend et al.). IL-1RA is a natural regulator of IL-1’s biological activity. It binds to the IL-1 receptors with similar affinity as IL-1, thereby inhibiting the proinflammatory activities of IL-1ɑ and IL-1ꞵ (Kinne et al.). In particular, mesenchymal stem cells have been shown to enhance tissue repair in an IL-1RA-dependent manner through the suppression of IL-1ꞵ production in dermal macrophages and enhanced expansion of immunosuppressive regulatory T cells in the skin (Harrell et al.). IL-1RA has also been shown to help in the treatment of rheumatoid arthritis (Cutolo), sepsis, asthma (Mao et al.), and inflammatory bowel diseases (Dosh et al.). This product is animal component-free.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité