Human Recombinant Galectin-1
Fournisseur: STEMCELL Technologies
100-0941
STMC100-0941EA 269 EUR InStock
STMC100-0941
Human Recombinant Galectin-1
Protéines et Peptides
Galectin-1 (Gal1) was the first characterized member of the galectin family of galactosidase-binding proteins, with over 15 mammalian galectins identified (Camby et al.; Salatino et al.). Gal1 comes in two forms: the oxidized monomer that acts as a cytokine, and the reduced dimer that acts as a lectin (Gaudet et al.). This product is in the dimer form. This cytokine is expressed in many tissues and has an immunosuppressive role in affecting T cell homeostasis by various mechanisms such as regulating apoptosis, cytokine secretion, cell adhesion, cell proliferation, and other effects (Camby et al.; Garín et al.; Gaudet et al.; Salatino et al.). In addition, Gal1 is thought to also play a role in axonal regeneration after injuries (Camby et al.; Garín et al.; Gaudet et al.; Salatino et al.). There are several therapeutic applications suggested for Gal1; overexpression has been suggested as a therapy for autoimmune and inflammatory diseases and enhancing axonal regeneration in injured nerves (Camby et al.; Gaudet et al.). In contrast, inhibition of Gal1 has been suggested to prevent tumor metastasis and cancer progression, as it may aid in cell adhesion, migration, and immune escape of cancer cells (Camby et al.).
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité