ImmunoCult™-XF Medium
Fournisseur: STEMCELL Technologies
100-0956
STMC100-0956EA 225 EUR InStock
STMC100-0956
ImmunoCult™-XF Medium
Milieux de culture cellulaire
cGMP, serum-free, and xeno-free medium for the expansion of human T cells.
Support the culture of human T cells and enhance your cell therapy development by using high-performance, cGMP-manufactured ImmunoCult™-XF medium. Designed for robust expansion for use in clinical applications without the need for serum or serum derivatives, this medium is also phenol red-free, xeno-free and has no added cytokines - providing complete flexibility for workflows. T cells can be harvested after just 10 to 12 days of culture and are able to produce cytokines, including interferon-gamma (IFN-γ) and IL4, upon restimulation. This ready-to-use medium is produced according to current GMP guidelines, ensuring the highest quality and consistency for reproducible results. ImmunoCult™-XF can be used individually or in combination with ImmunoCult™ Human CD3/CD28 T Cell Activator or ImmunoCult™ Human CD3/CD28/CD2 T Cell Activator for a gentle, bead-free activation of T cells.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité