Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Mitochondrial Tracking Dye, Deep Red

Fournisseur: STEMCELL Technologies

100-0995
STMC100-0995EA 247 EUR InStock
STMC100-0995
Mitochondrial Tracking Dye, Deep Red
Tests Tests cellulaires
Fluorescent hydrophobic dye that selectively accumulates in mitochondria - 500 tests.

  • Excitation wavelength: 640 nm
  • Emission wavelength: 659 nm
  • Easily crosses live cell membranes due to hydrophobicity
  • Partially dependent on mitochondrial membrane potential
  • Cell-retaining group produces longer retention times
  • Verified for use in fluorescence microscopy

Efficiently label mitochondria in proliferating and non-proliferating cells with Mitochondrial Tracking Dye, Deep Red. This fixable fluorescent dye uses a quick staining protocol that is well suited for high-throughput screening and is compatible with fluorescence microscopy. Mitochondrial Tracking Dye, Deep Red uses a labeling protocol with minimal hands-on time while promoting photostability and long retention times for enhanced staining efficiency.

Mitochondrial Tracking Dye, Deep Red is a hydrophobic dye that accumulates in mitochondria and is partially dependent on mitochondrial membrane potential. Upon depolarization, the dye relocalizes to the cytoplasm and becomes non-specific, but still retains its red fluorescence. Mitochondrial tracking dyes contain thiol chloromethyl, a cell-retaining group (Clutton et al. Cytometry A, 2019). The chloromethyl group increases staining efficiency by retaining the fluorescent dye in the mitochondria long after fixation (Clutton et al. Cytometry A, 2019). Due to its vivid far red fluorescence, Mitochondrial Tracking Dye, Deep Red is suitable for multi-labeling experiments such as microplate assays and flow cytometry. Use Mitochondrial Tracking dyes to quantify mitochondrial activity, localization, and abundance.

CAS. number: 67-68-5

Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').


Commander maintenant

 

COMMANDE

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité