Mitochondrial Tracking Dye, Blue
Fournisseur: STEMCELL Technologies
100-0996
STMC100-0996EA 256 EUR InStock
STMC100-0996
Mitochondrial Tracking Dye, Blue
Tests Tests cellulaires
Fluorescent hydrophobic dye that selectively accumulates in mitochondria - 500 tests.
  • Excitation wavelength: 344 nm
  • Emission wavelength: 469 nm
  • Easily crosses live cell membranes due to hydrophobicity
  • Partially dependent on mitochondrial membrane potential
  • Cell-retaining group produces longer retention times
Rapidly label mitochondria in proliferating and non-proliferating cells with Mitochondrial Tracking Dye, Blue. This fixable fluorescent dye uses a quick and simple staining protocol that is well suited for high-throughput screening and is compatible with fluorescence microscopy. Mitochondrial Tracking Dye, Blue uses a labeling protocol with minimal hands-on time while promoting photostability and long retention times for enhanced staining efficiency.
Mitochondrial Tracking Dye, Blue is a hydrophobic dye that accumulates in mitochondria and is partially dependent on mitochondrial membrane potential. Upon depolarisation, the dye relocalises to the cytoplasm and becomes non-specific, but still retains its blue fluorescence. This mitochondrial tracking dye contains a cell-retaining group thiol chloromethyl that increases staining efficiency by retaining fluorescence long after fixation (Clutton et al. Cytometry A, 2019). Mitochondrial Tracking Dye, Blue is suitable for multi-labeling experiments such as microplate assays and flow cytometry. Use mitochondrial tracking dyes to quantify mitochondrial activity, localisation, and abundance.
CAS No: 67-68-5
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Collapse All
Désignation Cdt Numéro de catalogue VWR Quantité
Mitochondrial tracking dye, blue 500 STMC100-0996
claquement
Veuillez sélectionner l'article
Entrer les détails
SPÉCIFICATIONS
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité