ImmunoCult™ Mouse B Cell Expansion Kit
Fournisseur: STEMCELL Technologies
100-1003
STMC100-1003EA 846 EUR InStock
STMC100-1003
ImmunoCult™ Mouse B Cell Expansion Kit
Milieux de culture cellulaire
Serum-free culture kit for in vitro expansion of mouse B cells.
Consistently expand mouse B cells in serum-free conditions without the use of problematic feeder cells. Use ImmunoCult™ Mouse B Cell Expansion Kit to provide optimised culture conditions for expanding B cells from mouse splenocytes with high yields. This kit comprises ImmunoCult™-XF B Cell Base Medium and ImmunoCult™ACF Mouse B Cell Expansion Supplement, providing a robust and easy-to-use culture system. After just 6 - 9 days in culture, cells can be harvested and used directly in downstream applications.
This kit is compatible with our other upstream and downstream products. For example, isolate B cells with EasySep™ Mouse Pan-B Cell Isolation Kit and expand them immediately using ImmunoCult™ Mouse B Cell Expansion Kit.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité