Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-Cytokeratin 20 Rabbit Polyclonal Antibody

Fournisseur: STEMCELL Technologies
100-1064
STMC100-1064EA 374 EUR InStock
STMC100-1064
Anti-Cytokeratin 20 Rabbit Polyclonal Antibody
Anticorps
Rabbit polyclonal antibody against human, mouse, rat cytokeratin 20.

  • Intestinal cells

This rabbit polyclonal antibody reacts with human, mouse, and rat cytokeratin 20, a low molecular weight, type I cytokeratin that is a major component of mature enterocytes and goblet cells. Cytokeratin 20 is expressed primarily in the gastrointestinal (GI) epithelium, urothelium, and Merkel cells and serves as a marker of GI differentiation. Use this polyclonal antibody in applications requiring detection of cytokeratin 20, such as western blotting (WB) assays, immunohistochemistry protocols (IHC-P), or immunofluorescence (IF). It is verified for use with immunocytochemistry (ICC). The cytokeratin 20 protein is responsible for the structural integrity of epithelial cells and maintains the intermediate filament organization in the intestinal epithelia. Cytokeratin 20 is also a useful marker of pancreatic and colorectal cancer and has been detected in adenocarcinomas of the colon, pancreas, stomach and biliary tract. Expression of cytokeratin 20 has also been associated with the progression and metastasis of breast cancer, suggesting that it may be a biomarker in predicting breast cancer prognosis.

Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: Cytokeratin 20
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité