EasySep™ Human EGFR Positive Selection Kit
Fournisseur: STEMCELL Technologies
100-1131
STMC100-1131EA 795 EUR InStock
STMC100-1131
EasySep™ Human EGFR Positive Selection Kit
Cell Separation Media
Immunomagnetic positive selection of human EGFR+ cells.
  • Fast and easy-to-use
  • Up to 98% purity
  • No columns required
Efficiently isolate cells expressing epidermal growth factor receptor , EGFR+ cells are labeled with antibodies and magnetic particles and isolated without the use of columns. Labeled cells remain in the tube while unlabeled cells are poured off. Depending on your research needs, either fraction may be used downstream.
This kit targets EGFR, which belongs to the ErbB family of tyrosine kinase receptors (RTK) expressed on the cell surface of circulating tumor cells (CTCs), macrophages, monocytes, plasma cells, and certain T cell subsets, such as effector CD4+ T cells and FoxP3-expressing regulatory CD4+ T cells (Tregs).
Attention: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité