Human Recombinant Resistin
Fournisseur: STEMCELL Technologies
100-1294
STMC100-1294EA 649 EUR InStock
STMC100-1294
Human Recombinant Resistin
Protéines et Peptides
Use resistin to modulate various cellular responses, such as promoting neutrophil extracellular trap (NET) formation (Jiang et al.) and regulating anti-inflammatory signaling pathways through toll-like receptor 4 (TLR4) (Jang et al., 2017). A member of the resistin-like molecule (RELM) family, resistin is produced by adipocytes in mice, and has been implicated in insulin resistance, reducing glucose tolerance, and insulin sensitivity in vivo (Li et al.). In humans, resistin is expressed predominantly in leukocytes, modulating inflammation in numerous diseases (Jamaluddin et al.; Jang et al., 2015; Mantula et al.). Studies also show that resistin facilitates vascular endothelial growth factor (VEGF)-A-dependent angiogenesis in human chondrosarcoma cells, highlighting it as a promising target for chondrosarcoma angiogenesis (Chen et al.). For consistency and reproducibility across your applications, resistin from STEMCELL comes lyophilised with ≥95% purity, and endotoxin levels are verified to be ≤1,0 EU/μg protein.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité