Human Recombinant Lipocalin-2, His Tag
Fournisseur: STEMCELL Technologies
100-1295
STMC100-1295EA 775 EUR InStock
STMC100-1295
Human Recombinant Lipocalin-2, His Tag
Protéines et Peptides
Lipocalin-2 (LCN2) is a member of the lipocalin superfamily, which share a highly conserved fold containing an eight-stranded antiparallel beta barrel (Flower), and transport small hydrophobic molecules. Playing a role in innate immunity, Lipocalin-2 acts as a potent bacteriostatic reagent by sequestering iron-containing siderophores (Flo et al.). It is mainly expressed in activated neutrophils, but is also secreted by other immune cells in response to infection. Lipocalin-2 has been implicated in multiple cellular processes, such as differentiation (Yang et al.), migration (Du et al.), and apoptosis (Devireddy et al.). In vitro studies suggest that lipocalin-2 is a potential therapeutic target for certain cancers, including colorectal cancer (CRC; Zhang et al.) and chronic lymphocytic leukemia (Bauvois et al.). This protein product contains a His-residue tag at the carboxyl end of the polypeptide chain. For consistency and reproducibility across your applications, lipocalin-2 from STEMCELL comes lyophilised with ≥94% purity, and endotoxin levels are verified to be ≤1.0 EU/μg protein.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité