Human Recombinant C-Reactive Protein
Fournisseur: STEMCELL Technologies
100-1297
STMC100-1297EA 579 EUR InStock
STMC100-1297
Human Recombinant C-Reactive Protein
Protéines et Peptides
A member of the pentraxin family of proteins, C-reactive protein (CRP) plays important roles in host defenses, including regulating complement activation in vivo (Sjoberg et al.), decreasing nitric oxide production, and inhibiting angiogenesis in vitro (Verma et al., 2002). At specific concentrations, CRP directly inhibits endothelial progenitor cell (EPC) differentiation, survival, and function, and is thus implicated in the development of cardiovascular diseases (Verma et al., 2004). CRP is primarily synthesized by liver hepatocytes, but also by other immune cells, such as macrophages and lymphocytes (Sproston and Ashworth). The expression of this acute-phase protein is induced by IL-6 and IL-1 secretion of macrophages and T-cells during inflammation. For consistency and reproducibility across your applications, C-reactive protein from STEMCELL comes lyophilised with ≥87% purity, and endotoxin levels are verified to be ≤1,0 EU/μg protein.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité