Human Recombinant ASAM, His tag
Fournisseur: STEMCELL Technologies
100-1331
STMC100-1331EA 610 EUR InStock
STMC100-1331
Human Recombinant ASAM, His tag
Protéines et Peptides
Adipocyte-specific adhesion molecule (ASAM) may play a role in cell-cell adhesion as it is expressed in epithelial cells of various tissues and is associated with tight junctions (Raschberger et al., Sze et al.). This type 1 transmembrane protein belongs to the cortical thymocyte marker in the Xenopus (CTX) family within the immunoglobulin superfamily. Sequence alignment analysis has shown that ASAM shares conserved characteristics with other CTX-like proteins, including extracellular V-type and C2-type domains, a transmembrane region, and a cytoplasmic tail (Raschperger et al.). ASAM is also expressed in adipocytes, and was found to be upregulated in white adipose tissue (WAT) in a rodent model of obesity and type II diabetes (Eguchi et al.). This protein contains a His-residue tag at the carboxyl end of the polypeptide chain. For consistency and reproducibility across your applications, adipocyte-specific adhesion molecule from STEMCELL comes lyophilised with ≥94% purity, and is verified by LAL analysis to ensure endotoxin levels are ≤1,0 EU/μg protein.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité