Human Recombinant Fetuin B, His tag
Fournisseur: STEMCELL Technologies
100-1333
STMC100-1333EA 549 EUR InStock
STMC100-1333
Human Recombinant Fetuin B, His tag
Protéines et Peptides
Fetuin B is thought to have functions in female fertility, metabolism, and liver disease. A protease inhibitor belonging to the fetuin family of proteins, within the cystatin superfamily, fetuin b’s structure includes two cystatin domains (CY1 and CY2) connected by a linker, with a CPDCP-trunk and a C-terminal region (CTR) (Cuppari et al.). This plasma protein originates in the liver (Denecke et al.) and acts to inhibit the activity of ovastacin, a metalloprotease that causes the zona pellucida to harden, leading to female infertility (Karmilin et al.). Fetuin B has also been reported to increase apoptosis, and inhibit migration and invasion of prostate cancer cells, in in vitro and in vivo settings (Zhan et al.). Elevated serum fetuin B has been linked to non-alcoholic fatty liver disease (NAFLD) (Zhu et al.). This protein contains a His-residue tag at the carboxyl end of the polypeptide chain. For consistency and reproducibility across your applications, apolipoprotein H from STEMCELL comes lyophilised with ≥92% purity, and is verified by LAL analysis to ensure endotoxin levels are ≤1,0 EU/μg protein.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité