Mouse Recombinant IFN beta
Fournisseur: STEMCELL Technologies
100-1337
STMC100-1337EA 827 EUR InStock
STMC100-1337
Mouse Recombinant IFN beta
Protéines et Peptides
A cytokine belonging to the type 1 interferon family, Interferon beta (IFN beta) binds IFN alpha/beta receptors (IFNAR) that activate tyrosine kinases and initiate the interferon-induced Jak-STAT signaling pathway, which modulates many key immune processes (Smieja et al.). In an experimental model involving cardiac fibroblasts isolated from rats, IFN beta was found to induce both pro- and anti-inflammatory cytokines production by activating different STAT proteins (Bolivar et al.). The anti-inflammatory effects of IFN beta have been studied in the context of autoimmune disorders, and there are currently multiple approved IFN beta drugs for treatment of relapsing forms of multiple sclerosis (Filipi and Jack). IFN beta is produced by immune cells, including macrophages, and non-immune cells, such as fibroblasts and epithelial cells (Ivashkiv and Donalin). The crystal structure of IFN beta shares characteristics with other type I interferons. It comprises five alpha-helices with four of them forming a helix bundle, and one long and three shorter loops connecting the helices (Karpusas et al.). For consistency and reproducibility across your applications, interferon alpha 1 from STEMCELL comes lyophilised with ≥87% purity, specific activity EC50 ≤18 pg/mL, and LAL analysis verification ensuring endotoxin levels are ≤1,0 EU/μg protein.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité