Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-Synaptophysin (MRX96) Rabbit Monoclonal Antibody

Fournisseur: STEMCELL Technologies
100-1345
STMC100-1345EA 799 EUR InStock
STMC100-1345
Anti-Synaptophysin (MRX96) Rabbit Monoclonal Antibody
Anticorps
Rabbit monoclonal antibody against human Synaptophysin (MRX96).

  • Neural cells Psc derived
  • Neurons

This rabbit monoclonal antibody (clone 249) reacts with synaptophysin, a 38 kDa integral transmembrane protein possessing four transmembrane domains and a major small synaptic synaptic vesicle protein. Use this recombinant monoclonal antibody in applications requiring detection of synaptophysin, such as immunohistochemistry (IHC), immunocytochemistry (ICC), and immunofluorescence (IF). This antibody clone has been verified for labeling synaptophysin-positive puncta in hPSC-derived forebrain-type neurons generated with STEMdiff™ Forebrain Neuron Differentiation Kit (Catalog #08600) and STEMdiff™ Forebrain Neuron Maturation Kit (Catalog #08605). It interacts with a variety of different motor proteins and synaptic vesicles including dynamin I, myosin V, vesicular proton pump V-ATPase, and adaptor protein 1 (AP-1). Synaptophysin is extensively involved in vesicular functions such as protein sorting and priming, as well as synapse formation, exocytosis, and endocytosis. Synaptophysin is also a potent marker of axonal damage; synaptophysin was discovered colocalised with amyloid precursor protein (APP), a marker of axonal damage in patients with multiple sclerosis. Synaptophysin deletion in knock-out mice have been correlated with reduced spatial learning and impaired object novelty recognition.

Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: Synaptophysin (MRX96)
Clonality: Monoclonal
Clone: 249
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité