Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-CD154 (CD40L) Mouse Monoclonal Antibody [clone: 5C8]

Fournisseur: STEMCELL Technologies
100-1352
STMC100-1352EA 930 EUR InStock
STMC100-1352
Anti-CD154 (CD40L) Mouse Monoclonal Antibody [clone: 5C8]
Anticorps
Mouse monoclonal antibody against human CD154 (CD40L).

  • T cells

This mouse monoclonal antibody (clone 5C8, APC-conjugated) reacts with human CD154 (CD40L), a member of the TNF-α superfamily with a molecular weight of 39 kDa. CD154, also known as CD40L, gp39, TRAP, and T-BAM, is a type II transmembrane glycoprotein that is predominantly expressed on activated T-cells, but is also found in NK cells, mast cells, basophils, and eosinophils. CD154 (CD40L) binds to CD40, a costimulatory protein found on antigen-presenting cells, such as B cells, monocytes, dendritic cells, and fibroblasts. The CD154 (CD40)-CD40L interaction induces the activation and proliferation of B-cells, allowing for antibody production, isotype switching, and B-cell memory development. Use this monoclonal antibody in applications requiring detection of CD154/CD40L such as flow cytometry (FC). CD154 (CD40L) expression is upregulated in inflammatory bowel disease, suggesting its role in regulating pathogenic cytokine production in T-cells. In vitro studies also demonstrate that the binding of CD40 to its ligand induces endothelial cell activation in vitro, highlighting the immunopathogenic role that CD40 and its ligand plays. The CD154 (CD40L), clone 5C8 antibody has been reported to block the interaction and ligation of CD40 with its ligand.

Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: CD154 (CD40L)
Clonality: Monoclonal
Clone: 5C8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité