Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-CD137 (ILA) Mouse Monoclonal Antibody [clone: 4B4-1] (Biotin)

Fournisseur: STEMCELL Technologies
100-1354
STMC100-1354EA 699 EUR InStock
STMC100-1354
Anti-CD137 (ILA) Mouse Monoclonal Antibody [clone: 4B4-1] (Biotin)
Anticorps
Mouse monoclonal antibody against human, monkey CD137 (ILA).

  • Dendritic cells
  • Nk cells
  • T cells

This mouse monoclonal antibody (clone 4B4-1) reacts with human and monkey CD137, a type I membrane protein and member of the TNF receptor superfamily. Following activation, it is expressed on multiple immune cells such as natural killer (NK), T, and dendritic cells. CD137 plays a role in cytokine activation, preventing activation-induced cell death and it also plays a role in promoting the activity of cytotoxic T cells. CD137 deficiency in humans has been hypothesized to inhibit differentiation and function of T cells; CD137-deficient mice have been recorded to have impaired T cell cytotoxicity, proliferation, and survival. As an immune stimulator, CD137 can influence the tumor microenvironment by increasing the cytotoxicity of T and NK cells as well as their ability to infiltrate tumors. For these reasons, CD137 represents a strong candidate for cancer immunotherapy. Use this monoclonal antibody in applications requiring detection of CD137, such as flow cytometry (FC).

Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type: Anti-CD137 (ILA) Mouse Monoclonal Antibody [clone: 4B4-1] (Biotin)
Antigen: CD137 (ILA)
Clonality: Monoclonal
Clone: 4B4-1
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Monkey

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité