SARS-CoV-2 (Spike Protein) Omicron/B.1.1.529 WT Reference Peptide Pool
Fournisseur: STEMCELL Technologies
100-1383
STMC100-1383EA 399 EUR InStock
STMC100-1383
SARS-CoV-2 (Spike Protein) Omicron/B.1.1.529 WT Reference Peptide Pool
Protéines et Peptides
Stimulating T cells with SARS-CoV-2 (Spike Protein) Omicron/B.1.1.529 WT reference peptide pool releases downstream cytokines and upregulates activation markers, enabling antigen-specific T cells to be detected or isolated for analysis. This peptide pool is a lyophilised mixture of 82 peptides from the spike glycoprotein of the wild-type SARS-CoV-2 virus. Consisting of the wild-type peptides that correspond to the mutated portions of the spike protein found in the SARS-CoV-2 Omicron variant (B.1.1.529), this pool is recommended to be used as a reference for the SARS-CoV-2 (Spike Protein) Omicron/B.1.1.529 mutation peptide pool. Through interactions with the angiotensin-converting enzyme 2 (ACE2) receptor, the spike protein enables the virus to attach to the cell membrane, and plays a critical role in viral entry (Hoffmann et al.; Walls et al.). Viral peptide pools are useful for a broad range of applications, including vaccine development, immunological research, and diagnostic assay development.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité