EBV (LMP2A) Peptide Pool
Fournisseur: STEMCELL Technologies
100-1389
STMC100-1389EA 549 EUR InStock
STMC100-1389
EBV (LMP2A) Peptide Pool
Protéines et Peptides
Stimulating T cells with EBV (LMP2A) peptide pool releases downstream cytokines and upregulates activation markers, enabling antigen-specific T cells to be detected or isolated for analysis. EBV (LMP2A) peptide pool is a lyophilised mixture of 27 peptides from latent membrane protein 2A (LMP2A) of Epstein-Barr virus (EBV; strain GD1), and consists of 15-mer peptides with 11-amino-acid overlaps that cover amino acids 1 to 118 on LMP2A. LMP2A contributes to the survival of EBV-transformed B cells (Mancao and Hammerschmidt) and mediates partial escape of infected B cells from recognition by EBV-specific CD8+ T cells (Rancan et al). Viral peptide pools are useful for a broad range of applications, including vaccine development, immunological research, and diagnostic assay development.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité