HPV16 (L1) Peptide Pool
Fournisseur: STEMCELL Technologies
100-1392
STMC100-1392EA 569 EUR InStock
STMC100-1392
HPV16 (L1) Peptide Pool
Protéines et Peptides
Stimulating T cells with HPV16 (L1) Peptide pool releases downstream cytokines and upregulates activation markers, enabling antigen-specific T cells to be detected or isolated for analysis. HPV16 (L1) Peptide pool is a lyophilised mixture of 124 peptides from major capsid protein L1 of human papillomavirus type 16 (HPV16) consisting of 15-mer peptides with 11-amino-acid overlaps that cover amino acids 1 to 505 on L1. L1 enters the nuclei of host cells via a Kap α2β1-mediated pathway (Nelson et al.). It cooperates with E2, another viral protein, to enhance viral DNA replication (Siddiqa et al.). L1 pentamers can assemble into virus-like particles (Chen et al. 2000; Chen et al. 2001). Viral peptide pools are useful for a broad range of applications, including vaccine development, immunological research, and diagnostic assay development.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité