HPV16 (L2) Peptide Pool
Fournisseur: STEMCELL Technologies
100-1393
STMC100-1393EA 589 EUR InStock
STMC100-1393
HPV16 (L2) Peptide Pool
Protéines et Peptides
Stimulating T cells with HPV16 (L2) peptide pool releases downstream cytokines and upregulates activation markers, enabling antigen-specific T cells to be detected or isolated for analysis. A lyophilised mixture of 116 peptides from minor capsid protein L2 of human papillomavirus type 16 (HPV16), HPV16 (L2) Peptide Pool consists of 15-mer peptides with 11-amino-acid overlaps that cover amino acids 1 to 473 on L2. L2 is equipped with multiple nuclear retention signals (Darshan et al.; Mamoor et al.), and enters the nuclei of host cells via several import pathways mediated by Kapα2β1 heterodimers, Kapβ2, and Kapβ3 (Darshan et al.). Viral peptide pools are useful for a broad range of applications, including vaccine development, immunological research, and diagnostic assay development.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité