HPV16 (E7) Peptide Pool
Fournisseur: STEMCELL Technologies
100-1394
STMC100-1394EA 524 EUR InStock
STMC100-1394
HPV16 (E7) Peptide Pool
Protéines et Peptides
Stimulating T cells with HPV16 (E7) Peptide pool releases downstream cytokines and upregulates activation markers, enabling antigen-specific T cells to be detected or isolated for analysis. HPV16 (E7) Peptide pool is a lyophilised mixture of 22 peptides from protein E7 of human papillomavirus type 16 (HPV16) consisting of 15-mer peptides with 11-amino-acid overlaps that cover amino acids 1 to 98 on protein E7. E7 is an oncoprotein that binds and mediates degradation of the retinoblastoma tumor suppressor protein pRB (Gonzalez et al.). E7 together with E6 demonstrated the ability to immortalize human keratinocytes in vitro, which likely has significance for HPVs oncogenic potential (Hawley-Nelson et al.). Viral peptide pools are useful for a broad range of applications, including vaccine development, immunological research, and diagnostic assay development.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité