HPV18 (L2) Peptide Pool
Fournisseur: STEMCELL Technologies
100-1397
STMC100-1397EA 569 EUR InStock
STMC100-1397
HPV18 (L2) Peptide Pool
Protéines et Peptides
Stimulating T cells with HPV18 (L2) peptide pool releases downstream cytokines and upregulates activation markers, enabling antigen-specific T cells to be detected or isolated for analysis. A lyophilised mixture of 113 peptides from minor capsid protein L2 of human papillomavirus type 18 (HPV18), HPV18 (L2) peptide pool consists of 15-mer peptides with 11-amino-acid overlaps that cover amino acids 1 to 462 on L2. With two nuclear localization signals, L2 may enter the nuclei of host cells via several import pathways mediated by Kap α2β1 heterodimers, Kap β2, and Kap β (Klucevsek et al.). L2 can also assemble into virus-like particles, and may have a role in encapsidation (Stauffer et al.). Viral peptide pools are useful for a broad range of applications, including vaccine development, immunological research, and diagnostic assay development.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité