JCV (VP1) Peptide Pool
Fournisseur: STEMCELL Technologies
100-1400
STMC100-1400EA 429 EUR InStock
STMC100-1400
JCV (VP1) Peptide Pool
Protéines et Peptides
Stimulating T cells with JCV (VP1) Peptide pool releases downstream cytokines and upregulates activation markers, enabling antigen-specific T cells to be detected or isolated for analysis. This peptide pool is a lyophilised mixture of 86 peptides from major capsid protein VP1 (VP1) of the JC polyomavirus (JCV), and consists of 15-mer peptides with 11-amino-acid overlaps that cover amino acids 1 to 354 on VP1. VP1 binds LSTc oligosaccharides found on host cell glycoproteins and glycolipids, and this interaction is said to be essential for JCV viral growth and infectivity (Neu et al.). The JCV VP1 (Del Valle et al.) and large T antigen (LT) (Delbue et al.) are detected in JCV-associated pathologies such as progressive multifocal leukoencephalopathy (PML) in patients with HIV. Viral peptide pools are useful for a broad range of applications, including vaccine development, immunological research, and diagnostic assay development.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité