HHV6 (U54) Peptide Pool
Fournisseur: STEMCELL Technologies
100-1401
STMC100-1401EA 569 EUR InStock
STMC100-1401
HHV6 (U54) Peptide Pool
Protéines et Peptides
Stimulating T cells with HHV6 (U54) peptide pool releases downstream cytokines and upregulates activation markers, enabling antigen-specific T cells to be detected or isolated for analysis. HHV6 (U54) peptide pool is a lyophilised mixture of 112 peptides from protein U54 of human herpesvirus 6B (HHV-6B; strain Z29), and consists of 15-mer peptides with 11-amino-acid overlaps that cover amino acids 1 to 459 on U54. U54 is a tegument transactivator and is suggested to be a homolog of the CMV UL82/83 (pp65/72K) protein (Isegawa et al.; Nicholas). In infected T cells, U54 has been shown to inhibit gene transcription of interleukin 2 (IL-2), the T cell growth factor, which likely contributes to viral persistence (Iampietro et al.). Viral peptide pools are useful for a broad range of applications, including vaccine development, immunological research, and diagnostic assay development.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité