VZV (IE63) Peptide Pool
Fournisseur: STEMCELL Technologies
100-1409
STMC100-1409EA 529 EUR InStock
STMC100-1409
VZV (IE63) Peptide Pool
Protéines et Peptides
Stimulating T cells with VZV (IE63) peptide pool releases downstream cytokines and upregulates activation markers, enabling antigen-specific T cells to be detected or isolated for analysis. VZV (IE63) peptide pool is a lyophilised mixture of 67 peptides from the transcriptional regulator ICP22 homolog (IE63) of Varicella-zoster virus (VZV; strain Oka vaccine), and consists of 15-mer peptides with 11-amino-acid overlaps that cover amino acids 1 to 278 on IE63. Expressed during VZV latency and reactivation (Lungu et al.), IE63 demonstrates repressive activity in both the cytoplasm and nucleus of infected cells (Bontems et al.). Host interferon alpha (IFN-α) activity is inhibited by VZV IE63, which likely contributes to VZV pathogenesis (Ambagala and Cohen). Viral peptide pools are useful for a broad range of applications, including vaccine development, immunological research, and diagnostic assay development.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité