Clostridium (Tetanus Toxin) Peptide Pool
Fournisseur: STEMCELL Technologies
100-1410
STMC100-1410EA 699 EUR InStock
STMC100-1410
Clostridium (Tetanus Toxin) Peptide Pool
Protéines et Peptides
Stimulating T cells with Clostridium (Tetanus Toxin) Peptide pool releases downstream cytokines and upregulates activation markers, enabling antigen-specific T cells to be detected or isolated for analysis. Clostridium Peptide pool is a lyophilised mixture of 326 peptides from tetanus toxin of Clostridium tetani (strain Massachusetts/E88), and consists of 15-mer peptides with 11-amino-acid overlaps that cover amino acids 1 to 1315 on tetanus toxin. Tetanus toxin entry into central neurons is mediated by the synaptic vesicle-binding protein SV2 (Yeh et al.), where it selectively cleaves synaptobrevin (Link et al.; Schiavo et al.). Viral peptide pools are useful for a broad range of applications, including vaccine development, immunological research, and diagnostic assay development.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité