CMV (IE1) Peptide Pool
Fournisseur: STEMCELL Technologies
100-1413
STMC100-1413EA 568 EUR InStock
STMC100-1413
CMV (IE1) Peptide Pool
Protéines et Peptides
Stimulating T cells with CMV (IE-1) peptide pool releases downstream cytokines and upregulates activation markers, enabling antigen-specific T cells to be detected or isolated for analysis. CMV (IE-1) peptide pool is a lyophilised mixture of 120 peptides from the immediate early protein IE1 (IE1) of human cytomegalovirus (CMV; strain AD169), and consists of 15-mer peptides with 11-amino-acid overlaps that cover amino acids 1 to 491 on IE1. CMV IE1 is a transcriptional transactivator and autoregulator (Cherrington and Mocarski). Studies show that IE1 promotes viral transcription by antagonizing histone deacetylation (Nevels et al.) and, along with IE2, may contribute to the survival of CMV-infected cells by blocking tumor necrosis factor alpha (TNF-α)-induced apoptosis (Zhu et al.). Viral peptide pools are useful for a broad range of applications, including vaccine development, immunological research, and diagnostic assay development.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité