SARS-CoV-2 (Spike Protein) Omicron BA.4/BA.5 Peptide Pool
Fournisseur: STEMCELL Technologies
100-1421
STMC100-1421EA 1655 EUR InStock
STMC100-1421
SARS-CoV-2 (Spike Protein) Omicron BA.4/BA.5 Peptide Pool
Protéines et Peptides
Stimulating T cells with SARS-CoV-2 (Spike Protein) Omicron BA.4/BA.5 Peptide pool releases downstream cytokines and upregulates activation markers, enabling antigen-specific T cells to be detected or isolated for analysis. This pool is provided as two lyophilised mixtures (subpools) from the spike glycoprotein of SARS-CoV-2 Omicron BA.4/BA.5 subvariant. The subpools contain 158 and 157 peptides, respectively, for a total of 315 peptides, and they consist of 15-mer peptides with 11-amino-acid overlaps that cover amino acids 1 to 1270 on the spike protein. Through interactions with the angiotensin-converting enzyme 2 (ACE2) receptor, the spike protein enables the virus to attach to the cell membrane, and plays a critical role in viral entry (Hoffmann et al.; Walls et al.). Viral peptide pools are useful for a broad range of applications, including vaccine development, immunological research, and diagnostic assay development.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité