Fixation Buffer
Fournisseur: STEMCELL Technologies
100-1450
STMC100-1450EA 79 EUR InStock
STMC100-1450
Fixation Buffer
Buffers Tampons de microscopie
Ideal for fixing cells prior to permeabilisation, fixation buffer can be used with Intracellular Permeabilisation Buffer (10X) to conduct intracellular staining for flow cytometry. This fixation buffer can also be used to preserve fluorescently labeled cells prior to analysis by flow cytometry.
Application: Immunology
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité