Intracellular Permeabilisation Buffer (10X)
Fournisseur: STEMCELL Technologies
100-1451
STMC100-1451EA 124 EUR InStock
STMC100-1451
Intracellular Permeabilisation Buffer (10X)
Buffers Tampons de microscopie
Ideal for the permeabilisation of fixed cells during labeling of cytokines and other cytoplasmic antigens. Intracellular Permeabilisation Buffer (10X) is uniquely formulated for optimal resolution and to reduce background and is suitable for use with fluorochrome-conjugated antibodies for flow cytometry.
Application: Immunology
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité