Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-TNF-alpha Mouse Monoclonal Antibody [clone: MT15B15] (Biotin)

Fournisseur: STEMCELL Technologies
100-1463
STMC100-1463EA 439 EUR InStock
STMC100-1463
Anti-TNF-alpha Mouse Monoclonal Antibody [clone: MT15B15] (Biotin)
Anticorps
Mouse monoclonal antibody against human TNF-alpha, biotin-conjugated.

  • Macrophages
  • T cells

This mouse monoclonal antibody (clone MT15B15) reacts with human tumor necrosis factor alpha (TNF-α). TNF-α is a pro-inflammatory cytokine primarily produced by macrophages and T cells in response to infection and inflammation. Initially generated as a ~26 kDa transmembrane precursor, TNF-α is then processed by metalloproteinases into a ~17 kDa soluble product. Both forms of TNF-α are active in trimeric form and exert their effects via binding to either TNFR1, present on the surface of nearly all human cell types, or TNFR2, which is localized to immune and endothelial cells. First described as having tumor-necrotic action, TNF-α signaling results in an inflammatory response that includes vasodilation and edema formation, adhesion of leukocytes to the epithelium, and blood coagulation. TNF-α also plays a key role in defense against bacterial, viral, and parasitic infections.

Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: TNF-alpha
Clonality: Monoclonal
Clone: MT15B15
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité