Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-CD80 (B7-1) Mouse Monoclonal Antibody [clone: 2D10.4] (PE)

Fournisseur: STEMCELL Technologies
100-1599
STMC100-1599EA 200 EUR InStock
STMC100-1599
Anti-CD80 (B7-1) Mouse Monoclonal Antibody [clone: 2D10.4] (PE)
Anticorps
Mouse monoclonal antibody against human, rhesus CD80 (B7-1), PE-conjugated.

  • B cells
  • Dendritic cells
  • Macrophages
  • T cells

This monoclonal antibody (clone 2D10.4) reacts with human cluster of differentiation 80 (CD80), also known as B7-1, a 60 kDa single chain type I transmembrane glycoprotein belonging to the immunoglobulin superfamily. CD80 is expressed by activated B cells, T cells, macrophages, and dendritic cells. Use this recombinant monoclonal antibody in applications requiring detection of CD80, such as flow cytometry (FC). This antibody clone has been verified for labeling CD80+ primary human M1 macrophages grown in ImmunoCult™-SF Macrophage Medium (Catalog #10961) containing Human Recombinant M-CSF (Catalog #78057), and then polarized with Human Recombinant IFN-gamma (Catalog #78020) and Lipopolysaccharide from E. coli (O55:B5) (Catalog #100-1270). CD80 has a high affinity for binding to two T cell receptors; CD28 and CD152 (also known as CTLA-4). The interaction of CD80 with CD28 provides a potent co-stimulatory signal for T cell activation through the CD3 complex, while its interaction with CD152 provides an inhibitory signal for T cell activation. CD80 together with CD86, another ligand of CD28 also known as B7-2, plays a major role in regulation of T cell activation. CD80 dysfunction is associated with diseases such as squamous cell carcinoma of gallbladder (SCC) and myocarditis. This antibody can also be used as a marker to assess classically activated type 1 (M1) macrophages.

Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: CD80 (B7-1)
Clonality: Monoclonal
Clone: 2D10.4
Conjugation: PE
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Rhesus monkey

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité