Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-CD4 Rat Monoclonal Antibody [clone: GK1.5] (FITC)

Fournisseur: STEMCELL Technologies
100-1603
STMC100-1603EA 43 EUR InStock
STMC100-1603
Anti-CD4 Rat Monoclonal Antibody [clone: GK1.5] (FITC)
Anticorps
Rat monoclonal antibody against mouse, hamster CD4, FITC-conjugated.

  • Dendritic cells
  • T cells
  • T cells CD4

This monoclonal antibody reacts with mouse cluster of differentiation 4 (CD4), a 55 kDa single chain type 1 transmembrane glycoprotein belonging to the immunoglobulin (Ig) superfamily. CD4 contains four extracellular Ig-like domains; D1 to D4. CD4 is expressed at relatively high levels by most thymocytes and a subpopulation of T cells; T helper/inducer cells, and at lower levels on dendritic cells. Use this recombinant monoclonal antibody in applications requiring detection of CD4, such as flow cytometry (FC), immunohistochemistry (IHC), and immunofluorescence (IF). Unlike humans, CD4 is not expressed by murine monocytes or macrophages. CD4 increases the affinity in the interaction between T cell receptor (TCR) and major histocompatibility complex II (MHC II) antigen complex by binding to a non-polymorphic region of MHC II and acting as a co-receptor to TCR in MHC II-restricted antigen recognition. CD4 also amplifies signals from TCR to the cytoplasm through the interaction of its intracellular domain with the cytoplasmic tyrosine kinases such as lymphocyte-specific protein tyrosine kinase (Lck). This monoclonal antibody is widely used as a phenotypic marker for CD4 expression. The RM4-5 clone can block binding of the GK1.5 antibody clone to CD4 on the cell surface. The GK1.5 antibody clone is also reported to be cross-reactive with Syrian hamster CD4.

Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type: Anti-CD4 Rat Monoclonal Antibody [clone: GK1.5] (FITC)
Antigen: CD4
Clonality: Monoclonal
Clone: GK1.5
Conjugation: FITC
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Hamster, Mouse

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité