Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-TCR Beta Hamster Monoclonal Antibody [clone: H57-597] (APC)

Fournisseur: STEMCELL Technologies
100-1611
STMC100-1611EA 151 EUR InStock
STMC100-1611
Anti-TCR Beta Hamster Monoclonal Antibody [clone: H57-597] (APC)
Anticorps
Hamster (Armenian) monoclonal antibody against mouse T cell receptor beta, APC-conjugated.

  • T cells

This monoclonal antibody (clone H57-597) reacts with the beta chain of mouse T cell receptor (TCR). TCR beta is a member of the immunoglobulin superfamily and together with the TCR alpha chain forms the alpha-beta TCR. TCR beta is expressed by thymocytes in a developmentally regulated manner and a majority of peripheral T cells. Small number of T cells express an alternative heteromer of gamma and delta protein chains, known as the gamma-delta TCR. TCR-CD3 complex in conjunction with co-receptors CD4 or CD8 enables the recognition and response to antigens attached to major histocompatibility complex (MHC) molecules on antigen-presenting cells. Crosslinking of the TCR complex with H57-597 induces activation and proliferation of T cells or apoptosis based on assay conditions. Use this recombinant monoclonal antibody in applications requiring detection of TCR beta, such as flow cytometry (FC), and the unconjugated antibody in Immunoperoxidase (IP), immunofluorescence (IF), Immunocytochemistry (ICC), Immunohistochemistry (IHC), and FC. The H57-597 antibody clone has been widely used as a phenotypic indicator for T cells expressing alpha-beta TCR. This antibody does not cross-react with cells expressing the gamma-delta TCR.

Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: TCR Beta
Clonality: Monoclonal
Clone: H57-597
Conjugation: APC
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité