Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-CD69 Hamster Monoclonal Antibody [clone: H1.2F3] (PE-Cy7)

Fournisseur: STEMCELL Technologies
100-1615
STMC100-1615EA 144 EUR InStock
STMC100-1615
Anti-CD69 Hamster Monoclonal Antibody [clone: H1.2F3] (PE-Cy7)
Anticorps
Hamster (Armenian) monoclonal antibody against mouse CD69, PE-Cy7-conjugated.

  • B cells
  • Macrophages
  • Monocytes
  • Nk cells
  • T cells

This monoclonal antibody (clone H1.2F3) reacts with mouse cluster of differentiation 69 (CD69), a type II transmembrane glycoprotein. CD69 is a 60 kDa disulfide bonded homodimer expressed on activated T cells, B cells, NK cells, neutrophils, and monocytes, where induction of CD69 rapidly occurs upon activation. CD69 is constitutively expressed on platelets and a subset of thymocytes. It functions as a co-stimulatory molecule that plays a role in activating and promoting the proliferation of T cells. Additionally, it can serve as an indicator for thymocytes engaged in T cell receptor (TCR)-mediated positive selection. Furthermore, the protein may act to transmit signals in natural killer cells and platelets. It has also been documented that co-stimulation with the H1.2F3 antibody clone enhances the activation of T cells and macrophages. CD69 dysfunction is associated with diseases such as coccidioidomycosis and asthma. Use this recombinant monoclonal antibody in applications requiring detection of CD69, such as flow cytometry (FC).

Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: CD69
Clonality: Monoclonal
Clone: H1.2F3
Conjugation: PE-Cy7
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité