Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-CD127 Rat Monoclonal Antibody [clone: A7R34] (PE-Cy7)

Fournisseur: STEMCELL Technologies
100-1630
STMC100-1630EA 269 EUR InStock
STMC100-1630
Anti-CD127 Rat Monoclonal Antibody [clone: A7R34] (PE-Cy7)
Anticorps
Rat monoclonal antibody against mouse CD127, PE-Cy7-conjugated.

  • B cells
  • Myeloid cells
  • T cells

This monoclonal antibody (clone A7R34) reacts with mouse cluster of differentiation 127 (CD127), a 60 - 90 kD type I transmembrane glycoprotein involved in the regulation of lymphopoiesis. CD127 is expressed on thymocytes, B cell precursors, T cells, and myeloid cells at the cell surface as a heterodimer with the common gamma chain (also known as CD132). This complex acts as a receptor for interleukin-7 (IL-7), which is an important cytokine in T and B cell development as well as in mature T cell homeostasis. Use this recombinant monoclonal antibody in applications requiring detection of CD127, such as flow cytometry (FC). Thymic stromal lymphopoietin (TSLP), a second cytokine, also binds to the receptor complex of CD127 and is involved in trigger activation of dendritic cells, allergy, and autoimmunity. CD127 has been reported to be a useful marker for identifying memory and effector T cells. The A7R34 antibody clone has been shown to block IL-7Rα signaling when administered in vivo.

Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: CD127
Clonality: Monoclonal
Clone: A7R34
Conjugation: PE-Cy7
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité