Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-KLRG1 Hamster Monoclonal Antibody [clone: 2F1] (PE)

Fournisseur: STEMCELL Technologies
100-1631
STMC100-1631EA 232 EUR InStock
STMC100-1631
Anti-KLRG1 Hamster Monoclonal Antibody [clone: 2F1] (PE)
Anticorps
Hamster (Syrian) monoclonal antibody against mouse KLRG1, PE-conjugated.

  • Nk cells
  • T cells CD8

This monoclonal antibody (clone 2F1) reacts with the mouse killer cell lectin-like receptor G1 (KLRG1), a 30 - 38 kDa homodimer receptor and a member of the lectin-like type 2 transmembrane receptor family of proteins. This receptor is expressed by activated, mature natural killer (NK) cells and by effector and memory T cells. KLRG1 expression has been associated with reduced proliferative capacity of activated T lymphocytes or reduced effector functions of activated NK cells. It has also been noted that cell surface expression of KLRG1 is upregulated by expression of the major histocompatibility (MHC) class I molecules, and this can be mediated by interactions with class I-specific Ly49 inhibitory receptors. The expression of mouse KLRG1, using 2F1 antibody clone, has not been detected on the surface of mouse mast cell lines, bone marrow-derived mast cells, or peritoneal mast cells. Use this recombinant monoclonal antibody in applications requiring detection of KLRG1, such as flow cytometry (FC).

Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: KLRG1
Clonality: Monoclonal
Clone: 2F1
Conjugation: PE
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité