ImmunoCult™ Mouse Treg Differentiation Supplement
Fournisseur: STEMCELL Technologies
10957
STMC10957EA 615 EUR InStock
STMC10957
ImmunoCult™ Mouse Treg Differentiation Supplement
Milieux de culture cellulaire
Supplement for the differentiation of mouse naïve CD4+ T cells into regulatory T cells (Tregs).
ImmunoCult™ mouse treg differentiation supplement contains a combination of recombinant mouse and human cytokines and a small molecule optimised to promote the differentiation of mouse naïve CD4+ T cells into regulatory T cells (Tregs). This supplement is intended for use with RPMI 1640 medium containing fetal bovine serum and other additives, as well as anti-mouse CD3 and anti-mouse CD28 monoclonal antibodies as activating agents.
This product is recommended for:
Research into the regulation of mouse naïve T cell differentiation and Treg function.
Development of procedures to expand and polarise naïve T cells toward a treg subset.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité