ImmunoCult™-SF Macrophage Medium
Fournisseur: STEMCELL Technologies
10961
STMC10961EA 195 EUR InStock
STMC10961
ImmunoCult™-SF Macrophage Medium
Milieux de culture cellulaire
Serum-free medium for differentiation of human monocytes to macrophages.
ImmunoCult™-SF Macrophage Medium has been developed for the in vitro culture and differentiation of human monocytes into macrophages when the appropriate cytokines and stimuli are added. The factors for differentiation and activation of macrophages have not been added to ImmunoCult™-SF Macrophage Medium. This provides users the flexibility to prepare a medium that meets their requirements. The medium is a specialised serum-free culture medium that can be used to differentiate human monocytes into M1 (classically activated) and M2a (alternatively activated) macrophages in a 6- or 8-day culture period.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité