ImmunoCult™-XF T Cell Expansion Medium
Fournisseur: STEMCELL Technologies
10981
STMC10981EA 225 EUR InStock
STMC10981
ImmunoCult™-XF T Cell Expansion Medium
Milieux de culture cellulaire
Serum-free and xeno-free medium for the expansion of human T cells.
ImmunoCult™-XF T Cell Expansion Medium is a serum-free and xeno-free medium optimised for the in vitro culture and expansion of human T cells isolated from peripheral blood. Recombinant cytokines, required for the optimal growth and expansion of T cells, have not been added to ImmunoCult™-XF T Cell Expansion Medium. This allows users the flexibility to prepare medium that meets their requirements.
This product is designed for cell therapy research applications following the recommendations of USP on Ancillary Materials, and we can currently work with you to qualify this reagent under an approved investigational new drug (IND) or clinical trial application (CTA).
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité