ImmunoCult™ Dendritic Cell Culture Kit
Fournisseur: STEMCELL Technologies
10985
STMC10985EA 509 EUR InStock
STMC10985
ImmunoCult™ Dendritic Cell Culture Kit
Milieux de culture cellulaire
For differentiation of human monocytes into dendritic cells.
Culture and differentiate human monocytes into dendritic cells (DCs) using the animal component-free (ACF) ImmunoCult™ dendritic cell culture kit.
For your convenience, the kit includes all the necessary components for you to generate active mature DCs from human monocytes in 7 days:
ImmunoCult™-ACF dendritic cell medium for the in vitro culture and differentiation of human monocytes into DCs.
ImmunoCult™-ACF dendritic cell differentiation Supplement to support the differentiation of immature DCs from human monocytes.
ImmunoCult™ dendritic cell maturation Supplement to support the maturation of immature human DCs.
For more information on protocols for culture and differentiation using ImmunoCult™ dendritic cell culture kit, please explore the Product Information Sheet (PIS).
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité