RosetteSep™ Complete Human Cord Blood Progenitor Enrichment Kit
Fournisseur: STEMCELL Technologies
15276
STMC15276EA 673 EUR InStock
STMC15276
RosetteSep™ Complete Human Cord Blood Progenitor Enrichment Kit
Cell Separation Media
Includes RosetteSep™ human cord blood progenitor kit and HetaSep™.
  • Fast and easy-to-use
  • Requires no special equipment or training
  • Untouched, viable cells
The RosetteSep™ Human Cord Blood Progenitor Cell Enrichment Cocktail with HetaSep™ is designed to isolate progenitor cells from cord blood by negative selection. The HetaSep™ minimises the required volume of the RosetteSep™ cocktail reagent by first concentrating the sample by sedimentation with HetaSep™ (STMC07906), a solution of hetastarch. The hematopoietic progenitors in the resultant buffy coat are then enriched using a modified RosetteSep™ protocol that uses less RosetteSep™ cocktail than is required for an unconcentrated sample. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognising CD2, CD3, CD14, CD16, CD19, CD24, CD56, CD61, CD66b and glycophorin A on red blood cells (RBCs). When centrifuged over a density gradient medium the unwanted cells pellet along with the RBCs. The purified progenitor cells are present as a highly enriched population at the interface between the plasma and the density gradient medium.
We regret to inform our customers that we can no longer include Ficoll-Paque™ Plus with this product. For performing cell isolation by density gradient centrifugation, we recommend purchasing Lymphoprep™ (STMC07801), which is available in a variety of sizes and formats. As with Ficoll-Paque™, Lymphoprep™ has a density of 1.077 g/mL, and after centrifugation, mononuclear cells remain at the plasma-Lymphoprep™ interface while granulocytes and erythrocytes sediment below.
Attention: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité