RosetteSep™ DM-L Density Medium
Fournisseur: STEMCELL Technologies
15705
STMC15705EA 101 EUR InStock
STMC15705
RosetteSep™ DM-L Density Medium
Cell Separation Media
Density gradient medium for use with RosetteSep™ CD3+ T cell, CD4+ T cell, CD8+ T cell, and B cell enrichment cocktails.
  • RosetteSep™ DM-L has a density of 1,081 g/ml
  • For use in cell isolation, In Vitro diagnostic applications
RosetteSep™ DM-L is a density gradient medium designed specifically for use with RosetteSep™ cocktails for the enrichment of specific human lymphocyte populations (CD3+ T cells, CD4+ T cells, CD8+ T cells, and B cells) from whole blood.
Attention: In light of the new EU Regulation 2017/746 (IVDR) of the European Parliament and of the Council on in vitro diagnostic medical devices, which is expected to become effective in 2022, STEMCELL recently deregistered and removed the CE IVD claims associated with these products, and they are now labelled 'For Research Use Only'. Please note there have been no changes to the form, function and quality assurances associated with these products. For more information please contact the STEMCELL Technologies Quality Assurance and Regulatory Department.
Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité