EasySep™ Dead Cell Removal (Annexin V) Kit
Fournisseur: STEMCELL Technologies
17899
STMC17899EA 367 EUR InStock
STMC17899
EasySep™ Dead Cell Removal (Annexin V) Kit
Milieux de séparation cellulaire
Immunomagnetic depletion of apoptotic (Annexin V+) cells from culture or tissue preparations.
  • Fast and easy-to-use
  • No columns required
  • Compatible across EasySep™, 'The Big Easy', and EasyEights™ platforms
The EasySep™ Dead Cell Removal (Annexin V) kit is designed to deplete apoptotic (Annexin V+) cells from cell culture or tissue preparations by immunomagnetic negative selection. Unwanted cells are labelled with Annexin V, antibodies and magnetic particles, and separated without columns using an EasySep™ magnet. labelled cells are separated using anMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité